CH6371379943 Chem Impex International, Inc.

1,2,3-Tribromopropane

  • MDL number MFCD00017884
  • Cas Number : 96-11-7
  • Molecular Weight : 280.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,2,3-tribromopropane
Inchi Code 1S/C3H5Br3/c4-1-3(6)2-5/h3H,1-2H2
Inchi Key FHCLGDLYRUPKAM-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Store at 0 - 8
Physical Form Clear, colorless to pale yellow liquid
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/1-2-3-tribromopropane/4657?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-2-3-tribromopropane/4657?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart