CH6371379951 Chem Impex International, Inc.

D-meta-Tyrosine

  • MDL number MFCD03788083
  • Cas Number : 32140-49-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-meta-Tyrosine
Purity 98
Storage temp. Store at 0-8
Physical Form Pale brown powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-meta-tyrosine/4665?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-meta-tyrosine/4665?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart