CH6371379952 Chem Impex International, Inc.

Boc-3,4-difluoro-DL-phenylalanine

  • MDL number MFCD03788084
  • Cas Number : 1214041-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103-110
Synonym Boc-3,4-difluoro-DL-phenylalanine
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-3-4-difluoro-dl-phenylalanine/4666?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-3-4-difluoro-dl-phenylalanine/4666?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart