CH6371379955 Chem Impex International, Inc.

Boc-3,4-Dimethoxy-D-Phenylalanine

  • MDL number MFCD01317725
  • Cas Number : 218457-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-D-Phe(3,4-Dimethoxy)-OH
Purity 98.0
Storage temp. Store at 0-8
Physical Form Pale white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-3-4-dimethoxy-d-phenylalanine/4676?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-3-4-dimethoxy-d-phenylalanine/4676?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart