CH6371379956 Chem Impex International, Inc.

Boc-3,4-dimethoxy-L-phenylalanine

  • MDL number MFCD01317724
  • Cas Number : 127095-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102 - 104 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-3-4-dimethoxy-l-phenylalanine/4677?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-3-4-dimethoxy-l-phenylalanine/4677?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart