CH6371379961 Chem Impex International, Inc.

Acetyl-4-iodo-DL-phenylalanine

  • MDL number MFCD03788088
  • Cas Number : 457615-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 193-202
Synonym Ac-DL-Phe(4-I)-OH
Purity 97
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-4-iodo-dl-phenylalanine/4683?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-4-iodo-dl-phenylalanine/4683?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart