CH6371379962 Chem Impex International, Inc.

Boc-allyl-D-glycine dicyclohexylammonium salt

  • MDL number MFCD01321012
  • Cas Number : 221352-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-D-Gly(allyl)-OH DCHA
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-allyl-d-glycine-dicyclohexylammonium-salt/4684?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-allyl-d-glycine-dicyclohexylammonium-salt/4684?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart