CH6371379974 Chem Impex International, Inc.

Boc-(R)-4-amino-6-methyl-heptanoic acid

  • MDL number MFCD01074530
  • Cas Number : 146453-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-gamma-L-leucine
Purity 98
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-r-i-4-amino-6-methyl-heptanoic-acid/4696?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-r-i-4-amino-6-methyl-heptanoic-acid/4696?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart