CH6371379977 Chem Impex International, Inc.

Boc-4-amino-1-methylpyrrole-2-carboxylic acid 1,2,3-benzotriazol-1-yl-ester

  • MDL number MFCD02179633
  • Cas Number : 77716-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-Py-OBt
Purity 99
Storage temp. Store at 0-8
Physical Form Amorphous white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-4-amino-1-methylpyrrole-2-carboxylic-acid-1-2-3-benzotriazol-1-yl-ester/4699?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-4-amino-1-methylpyrrole-2-carboxylic-acid-1-2-3-benzotriazol-1-yl-ester/4699?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart