CH6371379978 Chem Impex International, Inc.

Fmoc-(R,S)-3-amino-3-(1-naphthyl)propionic acid

  • MDL number MFCD02090642
  • Cas Number : 269078-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-DL-3-(naphthalene-1-yl)-3-amino-propionic acid
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-r-i-i-s-i-3-amino-3-1-naphthyl-propionic-acid/4700?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-r-i-i-s-i-3-amino-3-1-naphthyl-propionic-acid/4700?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart