CH6371379995 Chem Impex International, Inc.

Fmoc-homocyclohexyl-D-alanine

  • MDL number MFCD02179644
  • Cas Number : 269078-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-D-HomoCha-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-homocyclohexyl-d-alanine/4717?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-homocyclohexyl-d-alanine/4717?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart