CH6371379998 Chem Impex International, Inc.

Fmoc-(R,S)-Boc-imidazolidine-2-carboxylic acid

  • MDL number MFCD01321020
  • Cas Number : 207129-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-r-i-i-s-i-boc-imidazolidine-2-carboxylic-acid/4720?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-r-i-i-s-i-boc-imidazolidine-2-carboxylic-acid/4720?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart