CH6371380003 Chem Impex International, Inc.

Boc-(2S,4S)-4-phenylpyrrolidine-2-carboxylic acid

  • MDL number MFCD01321010
  • Cas Number : 96314-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-2-i-s-i-4-i-s-i-4-phenylpyrrolidine-2-carboxylic-acid/4725?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-2-i-s-i-4-i-s-i-4-phenylpyrrolidine-2-carboxylic-acid/4725?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart