CH6371380006 Chem Impex International, Inc.

Fmoc-3-styryl-D-alanine

  • MDL number MFCD01311775
  • Cas Number : 215190-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-D-Ala(styryl)-OH
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-styryl-d-alanine/4728?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-styryl-d-alanine/4728?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart