CH6371380008 Chem Impex International, Inc.

Fmoc-D-1,2,3,4-tetrahydronorharman-3-carboxylic acid

  • MDL number MFCD01861321
  • Cas Number : 268731-07-3
  • Molecular Weight : 438.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202-208°C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-d-1-2-3-4-tetrahydronorharman-3-carboxylic-acid/4730?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-d-1-2-3-4-tetrahydronorharman-3-carboxylic-acid/4730?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart