CH6371380014 Chem Impex International, Inc.

Fmoc-2-(2-aminoethoxy)-ethylamine hydrochloride

  • MDL number MFCD01321025
  • Cas Number : 221352-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-[2-(Fmoc-amino)ethoxy]ethylamine hydrochloride
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-2-2-aminoethoxy-ethylamine-hydrochloride/4736?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-2-2-aminoethoxy-ethylamine-hydrochloride/4736?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart