CH6371380023 Chem Impex International, Inc.

Boc-Leu-y(CH2NH)-Asp(OBzl)-OH

  • MDL number MFCD00273438
  • Cas Number : 204199-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form Amorphous white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-leu-y-ch-sub-2-sub-nh-asp-obzl-oh/4745?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-leu-y-ch-sub-2-sub-nh-asp-obzl-oh/4745?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart