CH6371380036 Chem Impex International, Inc.

3-Trifluoromethyl-D-phenylalanine

  • MDL number MFCD01860875
  • Cas Number : 14464-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Phe(3-CF3)-OH
Purity 0.98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-trifluoromethyl-d-phenylalanine/4758?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-trifluoromethyl-d-phenylalanine/4758?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart