CH6371380050 Chem Impex International, Inc.

Boc-3,4,5-trifluoro-D-phenylalanine

  • MDL number MFCD00797559
  • Cas Number : 205445-55-2
  • Molecular Weight : 319.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-3-4-5-trifluoro-d-phenylalanine/4772?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-3-4-5-trifluoro-d-phenylalanine/4772?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart