CH6371380052 Chem Impex International, Inc.

Fmoc-3,4,5-trifluoro-D-phenylalanine

  • MDL number MFCD00797584
  • Cas Number : 205526-31-4
  • Molecular Weight : 441.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166-172 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-4-5-trifluoro-d-phenylalanine/4774?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-4-5-trifluoro-d-phenylalanine/4774?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart