CH6371380074 Chem Impex International, Inc.

2-Methyl-L-phenylalanine

  • MDL number MFCD00044971
  • Cas Number : 80126-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Phe(2-Me)-OH
Purity 99.9
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-methyl-l-phenylalanine/4799?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-methyl-l-phenylalanine/4799?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart