CH6371380075 Chem Impex International, Inc.

2-Methyl-D-phenylalanine

  • MDL number MFCD01860870
  • Cas Number : 80126-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Phe(2-Me)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-methyl-d-phenylalanine/4800?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-methyl-d-phenylalanine/4800?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart