CH6371380076 Chem Impex International, Inc.

2-Bromo-L-phenylalanine

  • MDL number MFCD03092958
  • Cas Number : 42538-40-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Phe(2-Br)-OH
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-bromo-l-phenylalanine/4801?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-bromo-l-phenylalanine/4801?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart