CH6371380079 Chem Impex International, Inc.

3-Methyl-D-phenylalanine

  • MDL number MFCD01860871
  • Cas Number : 114926-39-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 226 - 231 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form Beige powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-methyl-d-phenylalanine/4804?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-methyl-d-phenylalanine/4804?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart