CH6371380080 Chem Impex International, Inc.

L-2,4-Dichlorophenylalanine

  • MDL number MFCD01860882
  • Cas Number : 111119-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Phe(2,4-DiCl)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-2-4-dichlorophenylalanine/4805?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-2-4-dichlorophenylalanine/4805?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart