CH6371380084 Chem Impex International, Inc.

2-Cyano-L-phenylalanine

  • MDL number MFCD01860884
  • Cas Number : 263396-42-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Phe(2-CN)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-cyano-l-phenylalanine/4809?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-cyano-l-phenylalanine/4809?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart