CH6371380085 Chem Impex International, Inc.

2-Cyano-D-phenylalanine

  • MDL number MFCD01860878
  • Cas Number : 263396-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point > 280 °C (dec.)
Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-cyano-d-phenylalanine/4810?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-cyano-d-phenylalanine/4810?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart