CH6371380087 Chem Impex International, Inc.

b-(3-Thienyl)-D-alanine

  • MDL number MFCD00065610
  • Cas Number : 152612-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-D-Ala(3-thienyl)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ß-3-thienyl-d-alanine/4812?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ß-3-thienyl-d-alanine/4812?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart