CH6371380091 Chem Impex International, Inc.

H-b-(2-Furyl)-D-alanine

  • MDL number MFCD01860879
  • Cas Number : 110772-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym H-beta-(2-Furyl)-D-alanine
Purity 0.98
Storage temp. Store at 0-8
Physical Form Off-white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/h-ß-2-furyl-d-alanine/4816?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/h-ß-2-furyl-d-alanine/4816?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart