CH6371380094 Chem Impex International, Inc.

Ac-Lys-D-Ala-D-Ala-OH

  • MDL number MFCD00214242
  • Cas Number : 28845-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Na-Acetyl-Lys-D-Ala-D-Ala-OH
Purity 97
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ac-lys-d-ala-d-ala-oh/4819?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ac-lys-d-ala-d-ala-oh/4819?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart