CH6371380095 Chem Impex International, Inc.

Z-Ala-His-OMe

  • MDL number MFCD00238403
  • Cas Number : 32303-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form White product
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-ala-his-ome/4820?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-ala-his-ome/4820?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart