CH6371380096 Chem Impex International, Inc.

Na-Z-L-asparagine methyl ester

  • MDL number MFCD02094371
  • Cas Number : 4668-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154-158 �C
Synonym Z-L-Asn-OMe
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-z-l-asparagine-methyl-ester/4822?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-z-l-asparagine-methyl-ester/4822?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart