CH6371380097 Chem Impex International, Inc.

Z-L-glutamic acid g-benzyl ester a-N-hydroxysuccinimide ester

  • MDL number MFCD02259504
  • Cas Number : 67413-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-Glu(OBzl)-OSu
Purity 95
Storage temp. Store at 0-8
Physical Form Colorless oil
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-glutamic-acid-gamma-benzyl-ester-alpha-i-n-i-hydroxysuccinimide-ester/4824?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-glutamic-acid-gamma-benzyl-ester-alpha-i-n-i-hydroxysuccinimide-ester/4824?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart