CH6371380099 Chem Impex International, Inc.

Z-glycyl glycine hydroxysuccinimide ester

  • MDL number MFCD00191098
  • Cas Number : 32943-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-Gly-Gly-OSu
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-glycyl-glycine-hydroxysuccinimide-ester/4826?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-glycyl-glycine-hydroxysuccinimide-ester/4826?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart