CH6371380101 Chem Impex International, Inc.

Z-N-Me-L-norleucine

  • MDL number MFCD02259505
  • Cas Number : 225386-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 37 - 41 °C
Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-i-n-i-me-l-norleucine/4831?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-i-n-i-me-l-norleucine/4831?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart