CH6371380102 Chem Impex International, Inc.

Z-N-methyl-L-threonine

  • MDL number MFCD02259506
  • Molecular Weight : 267.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-i-n-i-methyl-l-threonine/4832?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-i-n-i-methyl-l-threonine/4832?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart