CH6371380103 Chem Impex International, Inc.

Z-Pro-Gly-NH2

  • MDL number MFCD00238503
  • Cas Number : 35010-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-148
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-pro-gly-nh-sub-2-sub/4834?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-pro-gly-nh-sub-2-sub/4834?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart