CH6371380104 Chem Impex International, Inc.

Z-L-seryl-L-glycine ethyl ester

  • MDL number MFCD00191150
  • Cas Number : 4526-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White fine flowing powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-seryl-l-glycine-ethyl-ester/4835?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-seryl-l-glycine-ethyl-ester/4835?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart