CH6371380105 Chem Impex International, Inc.

Z-Ser-Thr-OMe

  • MDL number MFCD00191152
  • Cas Number : 7412-61-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-ser-thr-ome/4836?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-ser-thr-ome/4836?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart