CH6371380111 Chem Impex International, Inc.

Boc-Ala-Gly-Gly-OH

  • MDL number MFCD00078974
  • Cas Number : 115035-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-ala-gly-gly-oh/19719?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-ala-gly-gly-oh/19719?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart