CH6371380113 Chem Impex International, Inc.

Boc-Gly-Gly-Gly-OH

  • MDL number MFCD00190687
  • Cas Number : 28320-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-152 �C
Synonym Nalpha-t-Butoxycarbonyl-glycyl-glycyl-glycine
Purity 97.5
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-gly-gly-gly-oh/4855?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-gly-gly-gly-oh/4855?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart