CH6371380114 Chem Impex International, Inc.

Boc-L-trans-4-hydroxyproline tert-butyl ester

  • MDL number MFCD01318729
  • Cas Number : 170850-75-6
  • Molecular Weight : 287.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Physical Form White to off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-i-trans-i-4-hydroxyproline-i-tert-i-butyl-ester/4856?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-i-trans-i-4-hydroxyproline-i-tert-i-butyl-ester/4856?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart