CH6371380117 Chem Impex International, Inc.

Boc-N-methyl-O-benzyl-D-tyrosine

  • MDL number MFCD02259475
  • Cas Number : 138774-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-N-Me-D-Tyr(Bzl)-OH
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-n-i-methyl-i-o-i-benzyl-d-tyrosine/4861?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-n-i-methyl-i-o-i-benzyl-d-tyrosine/4861?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart