CH6371380123 Chem Impex International, Inc.

2-(4-Biphenylyl)isopropyloxycarbonyl-O-tert-butyl-L-threonine N-hydroxysuccinimide ester

  • MDL number MFCD00236886
  • Cas Number : 62020-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147-151
Synonym Bpoc-L-Thr(tBu)-OSu
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-4-biphenylyl-isopropyloxycarbonyl-i-o-i-i-tert-i-butyl-l-threonine-i-n-i-hydroxysuccinimide-ester/4879?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-4-biphenylyl-isopropyloxycarbonyl-i-o-i-i-tert-i-butyl-l-threonine-i-n-i-hydroxysuccinimide-ester/4879?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart