CH6371380125 Chem Impex International, Inc.

Bz-Tyr-4-Abz-OH

  • MDL number MFCD00055768
  • Cas Number : 37106-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Bentiromide
Purity 99
Storage temp. Store at 0-8
Physical Form Yellow powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/bz-tyr-4-abz-oh/4883?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/bz-tyr-4-abz-oh/4883?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart