CH6371380129 Chem Impex International, Inc.

Ac-D-phenylglycinol

  • MDL number MFCD00038782
  • Cas Number : 78761-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97-104 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form Off-whtite powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ac-d-phenylglycinol/4895?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ac-d-phenylglycinol/4895?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart