CH6371380130 Chem Impex International, Inc.

Dimethyl((3S)-4,4-dimethyl-3-(Boc-amino)-2-oxopentyl)phosphonate

  • MDL number MFCD00798804
  • Cas Number : 176504-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/dimethyl-3-i-s-i-4-4-dimethyl-3-boc-amino-2-oxopentyl-phosphonate/4899?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/dimethyl-3-i-s-i-4-4-dimethyl-3-boc-amino-2-oxopentyl-phosphonate/4899?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart