CH6371380136 Chem Impex International, Inc.

FA-Gly-Abu-NH2

  • MDL number MFCD00237636
  • Cas Number : 67607-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(2-Furyl)acryloyl-Gly-Abu-NH2
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fa-gly-abu-nh-sub-2-sub/4909?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fa-gly-abu-nh-sub-2-sub/4909?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart