CH6371380137 Chem Impex International, Inc.

Fmoc-D-allo-threoninol

  • MDL number MFCD01318736
  • Cas Number : 143143-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-144 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White product
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-d-i-allo-i-threoninol/4910?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-d-i-allo-i-threoninol/4910?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart