CH6371380139 Chem Impex International, Inc.

Fmoc-Gly-Gly-Gly-OH

  • MDL number MFCD00190881
  • Cas Number : 170941-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-glycyl-glycyl-glycine
Purity 97
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-gly-gly-gly-oh/4913?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-gly-gly-gly-oh/4913?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart